missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hidroxi-5-nitrobenzaldehído, 95 %, Thermo Scientific™

Catalog Number p-7034211
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
711022-47-8
C7H4FNO4
185.11
MFCD11846408
CZOMOYVUYOVNHX-UHFFFAOYSA-N
18452164
3-fluoro-4-hidroxi-5-nitrobenzaldehído
OC1=C(C=C(C=O)C=C1F)[N+]([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
711022-47-8
C7H4FNO4
185.11
MFCD11846408
CZOMOYVUYOVNHX-UHFFFAOYSA-N
18452164
3-fluoro-4-hidroxi-5-nitrobenzaldehído
OC1=C(C=C(C=O)C=C1F)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Fluoro-4-hydroxy-5-nitrobenzaldehyde
Cantidad 250 mg
Sensibilidad Air Sensitive
Formula Weight (peso de la fórmula) 185.11
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'