Learn More
3-Dimethylamino-1-(3-pyridyl)-2-propen-1-one, TRC
Product Code. 30371676
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30371676 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30371676 Supplier Toronto Research Chemicals Supplier No. TRCD462145500MG
 Restricted Item
Fecha estimada de envoi: 20-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Dimethylamino-1-(3-pyridyl)-2-propen-1-one
CAS 55314-16-4
Molecular Formula C10H12N2O
Quantity 500 mg
Synonym Imatinib Dimethylamino Impurity
InChI Formula InChI=1S/C10H12N2O/c1-12(2)7-5-10(13)9-4-3-6-11-8-9/h3-8H,1-2H3/b7-5+
SMILES CN(C)\C=C\C(=O)c1cccnc1
Molecular Weight (g/mol) 176.22