Learn More
Cloruro de 3-cloro-4-(trifluorometil)benzoilo, 97 %, Thermo Scientific™
Catalog Number p-7082733
Click to view available options
Cantidad:
1 g
5 g
1092460-77-9
C8H3Cl2F3O
243.006
MFCD11519325
KKXPAIGCYQCQHM-UHFFFAOYSA-N
3-chloro-4-trifluoromethyl benzoyl chloride, 3-chloro-4-trifluoromethyl benzoyl chloride, jrd
21275239
cloruro de 3-cloro-4-(trifluorometil)benzoilo
C1=CC(=C(C=C1C(=O)Cl)Cl)C(F)(F)F
1092460-77-9
C8H3Cl2F3O
243.006
MFCD11519325
KKXPAIGCYQCQHM-UHFFFAOYSA-N
3-chloro-4-trifluoromethyl benzoyl chloride, 3-chloro-4-trifluoromethyl benzoyl chloride, jrd
21275239
cloruro de 3-cloro-4-(trifluorometil)benzoilo
C1=CC(=C(C=C1C(=O)Cl)Cl)C(F)(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Chloro-4-(trifluoromethyl)benzoyl chloride
Color Incolora
Índice de refracción 1.503
Cantidad 1 g
Número UN UN3265
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 243.01
Porcentaje de pureza 97%
Forma física Líquido