missing translation for 'onlineSavingsMsg'
Learn More

3'-Cloro-2'-metilacetanilida, 98 %, Alfa Aesar™

Catalog Number p-7076518
Click to view available options
Cantidad:
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
7463-35-6
C9H10ClNO
183.635
MFCD00018264
NALGTKRTIJHBBK-UHFFFAOYSA-N
n-3-chloro-2-methylphenyl acetamide, 3-chloro-2-methylacetanilide, acetamide, n-3-chloro-2-methylphenyl, 3'-chloro-2'-methylacetanilide, 3-chloro-2-methyl-n-acetylaniline, 3'-chloro-o-acetotoluidide, 3'-chloro-ortho-acetotoluidide, n-acetyl-3-chloro-2-methylaniline, 2-acetamido-6-chlorotoluene, n-2-methyl-3-chlorophenyl acetamide
81969
N-(3-cloro-2-metilfenil)acetamida
CC1=C(C=CC=C1Cl)NC(=O)C
Este artículo no se puede devolver. Vea la política de devoluciones
7463-35-6
C9H10ClNO
183.635
MFCD00018264
NALGTKRTIJHBBK-UHFFFAOYSA-N
n-3-chloro-2-methylphenyl acetamide, 3-chloro-2-methylacetanilide, acetamide, n-3-chloro-2-methylphenyl, 3'-chloro-2'-methylacetanilide, 3-chloro-2-methyl-n-acetylaniline, 3'-chloro-o-acetotoluidide, 3'-chloro-ortho-acetotoluidide, n-acetyl-3-chloro-2-methylaniline, 2-acetamido-6-chlorotoluene, n-2-methyl-3-chlorophenyl acetamide
81969
N-(3-cloro-2-metilfenil)acetamida
CC1=C(C=CC=C1Cl)NC(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3′-Chloro-2′-methylacetanilide
Punto de fusión 155°C to 157°C
Envase Frasco de plástico
Cantidad 25 g
Formula Weight (peso de la fórmula) 183.64
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'