missing translation for 'onlineSavingsMsg'
Learn More

3-Cloro-2-metoxipiridina, 95 %, Thermo Scientific™

Catalog Number 11441800
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
13472-84-9
C6H6ClNO
143.57
MFCD03095227
MSICGKUDCSUMJZ-UHFFFAOYSA-N
3-chloro-2-methoxy-pyridine, pyridine, 3-chloro-2-methoxy, 2-methoxy-3-chloropyridine, pubchem1189, acmc-1c0pw, ksc494g7h, 3-chloranyl-2-methoxy-pyridine
13295749
3-cloro-2-metoxipiridina
COC1=C(C=CC=N1)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
13472-84-9
C6H6ClNO
143.57
MFCD03095227
MSICGKUDCSUMJZ-UHFFFAOYSA-N
3-chloro-2-methoxy-pyridine, pyridine, 3-chloro-2-methoxy, 2-methoxy-3-chloropyridine, pubchem1189, acmc-1c0pw, ksc494g7h, 3-chloranyl-2-methoxy-pyridine
13295749
3-cloro-2-metoxipiridina
COC1=C(C=CC=N1)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Chloro-2-methoxypyridine
Cantidad 1 g
Formula Weight (peso de la fórmula) 143.57
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'