missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldoxima, 97 %, Thermo Scientific™

Catalog Number 11440638
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
51873-95-1
C7H6BrNO
200.04
MFCD00143098
DBGBMIPNNIZQNN-UITAMQMPSA-N
3-bromobenzaldoxime, 3-bromobenzaldehyde oxime, e-3-bromobenzaldehyde oxime, pubchem3732, e-n-3-bromophenyl methylidene hydroxylamine, z-3-bromobenzaldehyde oxime, benzaldehyde, 3-bromo-, oxime, z-n-3-bromophenyl methylidene hydroxylamine, ne-n-3-bromophenyl methylidene hydroxylamine
5375574
O\N=C/C1=CC=CC(Br)=C1
Este artículo no se puede devolver. Vea la política de devoluciones
51873-95-1
C7H6BrNO
200.04
MFCD00143098
DBGBMIPNNIZQNN-UITAMQMPSA-N
3-bromobenzaldoxime, 3-bromobenzaldehyde oxime, e-3-bromobenzaldehyde oxime, pubchem3732, e-n-3-bromophenyl methylidene hydroxylamine, z-3-bromobenzaldehyde oxime, benzaldehyde, 3-bromo-, oxime, z-n-3-bromophenyl methylidene hydroxylamine, ne-n-3-bromophenyl methylidene hydroxylamine
5375574
O\N=C/C1=CC=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Bromobenzaldoxime
Punto de fusión 70°C to 74°C
Cantidad 5 g
Beilstein 2040559
Nombre IUPAC (NE)-N-[(3-bromofenil)metilideno]hidroxilamina
Formula Weight (peso de la fórmula) 200.03
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'