missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromoacetofenona dietil cetal, 96 %, estabilizado con carbonato de potasio, Thermo Scientific™

Catalog Number p-7031178
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
480439-43-8
C12H17BrO2
273.17
MFCD14636457
XKQAVDMOLWVSRW-UHFFFAOYSA-N
3'-bromoacetophenone diethyl ketal, 1-bromo-3-1,1-diethoxyethyl benzene, 3-bromoacetophenone diethyl ketal,
73995132
1-bromo-3-(1,1-dietoxietil)benceno
CCOC(C)(C1=CC(=CC=C1)Br)OCC
Este artículo no se puede devolver. Vea la política de devoluciones
480439-43-8
C12H17BrO2
273.17
MFCD14636457
XKQAVDMOLWVSRW-UHFFFAOYSA-N
3'-bromoacetophenone diethyl ketal, 1-bromo-3-1,1-diethoxyethyl benzene, 3-bromoacetophenone diethyl ketal,
73995132
1-bromo-3-(1,1-dietoxietil)benceno
CCOC(C)(C1=CC(=CC=C1)Br)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3'-Bromoacetophenone diethyl ketal
Nombre de nota stabilized with Potassium carbonate
Cantidad 5 g
Formula Weight (peso de la fórmula) 273.17
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'