missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-ácido nitrobenzoico, 99 %, Thermo Scientific™

Catalog Number p-7047690
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
6307-83-1
C7H4BrNO4
246.016
MFCD00100098
AXRKIZCFYZBBPX-UHFFFAOYSA-N
3-bromo-5-nitrobenzoic acid, 5-bromo-3-nitrobenzoic acid, benzoic acid, 3-bromo-5-nitro, 3-bromo-5-nitro-benzoic acid, 3-bromo-5-nitrobenzoicacid, pubchem4734, acmc-209ncw, ksc357s4t, 3-bromo-5-nitro benzoic acid
239336
C1=C(C=C(C=C1[N+](=O)[O-])Br)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
6307-83-1
C7H4BrNO4
246.016
MFCD00100098
AXRKIZCFYZBBPX-UHFFFAOYSA-N
3-bromo-5-nitrobenzoic acid, 5-bromo-3-nitrobenzoic acid, benzoic acid, 3-bromo-5-nitro, 3-bromo-5-nitro-benzoic acid, 3-bromo-5-nitrobenzoicacid, pubchem4734, acmc-209ncw, ksc357s4t, 3-bromo-5-nitro benzoic acid
239336
C1=C(C=C(C=C1[N+](=O)[O-])Br)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Bromo-5-nitrobenzoic acid
Punto de fusión 159°C to 161°C
Cantidad 250 mg
Beilstein 2051365
Formula Weight (peso de la fórmula) 246.02
Porcentaje de pureza 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'