Learn More
3-Bromo-4-hydroxybenzaldehyde, TRC
Product Code. 30338381
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30338381 5 g 5g
1 options
This item is not returnable. View return policy
Product Code. 30338381 Supplier Toronto Research Chemicals Supplier No. TRCB4153805G
 Restricted Item
Fecha estimada de envoi: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Bromo-4-hydroxybenzaldehyde
CAS 2973-78-6
Molecular Formula C7H5BrO2
Quantity 5 g
Synonym 2-Bromo-4-formylphenol, 4-Hydroxy-3-bromobenzaldehyde, NSC 220227
InChI Formula InChI=1S/C7H5BrO2/c8-6-3-5(4-9)1-2-7(6)10/h1-4,10H
SMILES Oc1ccc(C=O)cc1Br
IUPAC Name 3-bromo-4-hydroxybenzaldehyde
Molecular Weight (g/mol) 201.02
Formula Weight 199.9473