Learn More
Ácido 3-(Boc-amino)ciclohexanecarboxílico, +98 %, Thermo Scientific Chemicals
Catalog Number p-7034610
Click to view available options
Cantidad:
250 mg
1 g
5 g
334932-13-7
C12H21NO4
243.303
MFCD03453252
JSGHMGKJNZTKGF-UHFFFAOYSA-N
3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 3-boc-amino cyclohexanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, 3-2-methylpropan-2-yl oxycarbonylamino cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino, cis-3-boc-amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, acmc-209fp1
4420316
Ácido 3-[(2-metilpropan-2-il)oxicarbonilamino]ciclohexano-1-carboxílico
CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O
334932-13-7
C12H21NO4
243.303
MFCD03453252
JSGHMGKJNZTKGF-UHFFFAOYSA-N
3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 3-boc-amino cyclohexanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, 3-2-methylpropan-2-yl oxycarbonylamino cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino, cis-3-boc-amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, acmc-209fp1
4420316
Ácido 3-[(2-metilpropan-2-il)oxicarbonilamino]ciclohexano-1-carboxílico
CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O
3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El ácido 3(Boc-amino)-ciclohexanecarboxílico se utiliza como intermediario farmacéutico.
Solubilidad
Ligeramente soluble en agua (2,1 g/l a 25 °C).
Notas
Estables en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(Boc-amino)cyclohexanecarboxylic acid
Información de solubilidad Slightly soluble in water (2.1g/L at 25°C).
Formula Weight (peso de la fórmula) 243.3
Porcentaje de pureza ≥98%