missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzonitrilo, 99 %, Thermo Scientific Chemicals

Catalog Number p-7021595
Click to view available options
Cantidad:
10 g
50 g
Este artículo no se puede devolver. Vea la política de devoluciones
2237-30-1
C7H6N2
118.139
MFCD00007756
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
16702
3-aminobenzonitrilo
C1=CC(=CC(=C1)N)C#N
Este artículo no se puede devolver. Vea la política de devoluciones
2237-30-1
C7H6N2
118.139
MFCD00007756
NJXPYZHXZZCTNI-UHFFFAOYSA-N
3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
16702
3-aminobenzonitrilo
C1=CC(=CC(=C1)N)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-Aminobenzonitrile
Punto de fusión 51°C to 53°C
Punto de ebullición 290°C
Temperatura de inflamación >110°C (230°F)
Olor Almond-like
Cantidad 10 g
Número UN UN3439
Formula Weight (peso de la fórmula) 118.14
Porcentaje de pureza 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'