Learn More
3-Amino-2,5-dichlorobenzoic Acid, TRC
Product Code. 30324276
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30324276 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30324276 Supplier Toronto Research Chemicals Supplier No. TRCA6052731G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chloramben
CAS 133-90-4
Molecular Formula C7 H5 Cl2 N O2
Quantity 1 g
Synonym 3-Amino-2,5-dichlorobenzoic acid, 2,5-Dichloro-3-aminobenzoic acid, Ambiben, Amiben, Amibin, Amoben, Chloramben, Vegiben
InChI Formula InChI=1S/C7H5Cl2NO2/c8-3-1-4(7(11)12)6(9)5(10)2-3/h1-2H,10H2,(H,11,12)
SMILES Nc1cc(Cl)cc(C(=O)O)c1Cl
IUPAC Name 3-amino-2,5-dichlorobenzoic acid
Molecular Weight (g/mol) 206.03
Formula Weight 204.9697