missing translation for 'onlineSavingsMsg'
Learn More

3-(6-Tiocarbamoil-2-benzimidazolil)tiobenzamida, 97 %, Thermo Scientific™

Catalog Number p-7034462
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1256483-51-8
C15H12N4S2
312.409
MFCD16658841
XKUFJCIMNRMHRY-UHFFFAOYSA-N
3-6-thiocarbamoyl-2-benzimidazolyl thiobenzamide, 2-3-carbamothioylphenyl-3h-1,3-benzodiazole-5-carbothioamide
73995302
2-(3-carbamotioilfenil)-3H-benzimidazol-5-carbotioamida
C1=CC(=CC(=C1)C(=S)N)C2=NC3=C(N2)C=C(C=C3)C(=S)N
Este artículo no se puede devolver. Vea la política de devoluciones
1256483-51-8
C15H12N4S2
312.409
MFCD16658841
XKUFJCIMNRMHRY-UHFFFAOYSA-N
3-6-thiocarbamoyl-2-benzimidazolyl thiobenzamide, 2-3-carbamothioylphenyl-3h-1,3-benzodiazole-5-carbothioamide
73995302
2-(3-carbamotioilfenil)-3H-benzimidazol-5-carbotioamida
C1=CC(=CC(=C1)C(=S)N)C2=NC3=C(N2)C=C(C=C3)C(=S)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(6-Thiocarbamoyl-2-benzimidazolyl)thiobenzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 312.41
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'