Learn More
3,6-Dihydro-2H-pyridine-1-N-Boc-4-boronic Acid Pinacol Ester, TRC
Product Code. 30004423
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004423 100 mg 100mg
30003951 500 mg 500mg
30010946 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004423 Supplier Toronto Research Chemicals Supplier No. TRCD453413100MG
 Restricted Item
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 286961-14-6
Molecular Formula C16 H28 B N O4
Molecular Weight (g/mol) 309.209
Synonym tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydropyridine-1(2H)-carboxylate
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=CC1)B2OC(C)(C)C(C)(C)O2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C16H28BNO4/c1-14(2,3)20-13(19)18-10-8-12(9-11-18)17-21-15(4,5)16(6,7)22-17/h8H,9-11H2,1-7H3
Formula Weight 309.211
Chemical Name or Material 3,6-Dihydro-2H-pyridine-1-N-Boc-4-boronic Acid Pinacol Ester