missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoic acid, 98+%

Product Code. 11407827
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407827 1000 g 1000g
11417827 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11407827 Supplier Thermo Scientific Alfa Aesar Supplier No. A16502.0B
 Restricted Item
Fecha estimada de envoi: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol

3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.

Solubility
It is Soluble in ethanol (50 mg/mL), methanol, alcohol (very), and glacial acetic acid (very). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases, Strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-34-3
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007253
InChI Key VYWYYJYRVSBHJQ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
PubChem CID 7433
ChEBI CHEBI:73914
IUPAC Name 3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 204°C to 208°C
Density 1.68
Quantity 1000 g
UN Number UN1325
Beilstein 1914286
Merck Index 14,3276
Solubility Information It is Soluble in ethanol (50mg/mL),methanol,alcohol (very),and glacial acetic acid (very). Insoluble in water.
Formula Weight 212.12
Percent Purity ≥98%
Chemical Name or Material 3,5-Dinitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.