Learn More
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, TRC
Product Code. 30132857
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132857 50 g 50g
30010752 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132857 Supplier Toronto Research Chemicals Supplier No. TRCD49371550G
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1620-98-0
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.33
Synonym 2,6-Di-tert-Butyl-4-formylphenol, 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzaldehyde, 4-Formyl-2,6-di-tert-butylphenol, 4-Hydroxy-3,5-di-tert-butylbenzaldehyde, NSC 14450
SMILES CCOC(=O)C1CC2C(N2)C(OC(CC)CC)C=1
Specifications
Quantity 50 g
InChI Formula InChI=1S/C15H22O2/c1-14(2,3)11-7-10(9-16)8-12(13(11)17)15(4,5)6/h7-9,17H,1-6H3
Formula Weight 234.162
Percent Purity >95%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzaldehyde
Grade HPLC