missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluorometil)fenilhidracina, 97 %, Thermo Scientific™

Catalog Number p-7022275
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
886-35-1
C8H6F6N2
244.14
MFCD00051606
OULWACKZBGBRNT-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenylhydrazine, 3,5-bis-trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 3,5-di trifluoromethyl phenylhydrazine, 1-3,5-bis trifluoromethyl phenyl hydrazine, hydrazine, 3,5-bis trifluoromethyl phenyl, pubchem1080, pubchem3289, timtec-bb sbb028449
522901
[3,5-Bis(trifluorometil)fenil]hidracina
NNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
886-35-1
C8H6F6N2
244.14
MFCD00051606
OULWACKZBGBRNT-UHFFFAOYSA-N
3,5-bis trifluoromethyl phenylhydrazine, 3,5-bis-trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 3,5-di trifluoromethyl phenylhydrazine, 1-3,5-bis trifluoromethyl phenyl hydrazine, hydrazine, 3,5-bis trifluoromethyl phenyl, pubchem1080, pubchem3289, timtec-bb sbb028449
522901
[3,5-Bis(trifluorometil)fenil]hidracina
NNC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Aplicaciones
La 3,5-bis(trifluorometil)fenilhidracina se utiliza para medir la concentración de formaldehído.

Solubilidad
Poco soluble en agua.

Notas
Almacenar lejos de los agentes oxidantes fuertes. Mantener el recipiente herméticamente cerrado. Almacénese en un lugar fresco y seco en recipientes herméticos.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3,5-Bis(trifluoromethyl)phenylhydrazine
Punto de fusión 80°C to 83°C
Cantidad 1 g
Beilstein 1843213
Información de solubilidad Sparingly Soluble is water.
Formula Weight (peso de la fórmula) 244.14
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'