missing translation for 'onlineSavingsMsg'
Learn More

3,5-bis(Trifluorometil)bencilamina, 97 %, Thermo Scientific™

Catalog Number p-7027643
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
85068-29-7
C9H7F6N
243.152
MFCD00009909
DHVHORCFFOSRBP-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
521099
[3,5-bis(trifluorometil)fenil]metanamina
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN
Este artículo no se puede devolver. Vea la política de devoluciones
85068-29-7
C9H7F6N
243.152
MFCD00009909
DHVHORCFFOSRBP-UHFFFAOYSA-N
3,5-bis trifluoromethyl benzylamine, 3,5-bis trifluoromethyl phenyl methanamine, 3,5-di trifluoromethyl benzylamine, 3,5-ditrifluoromethylbenzylamine, mbt-bym, 3,5-bis trifluoromethyl phenyl methylamine, benzenemethanamine, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzylamine
521099
[3,5-bis(trifluorometil)fenil]metanamina
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3,5-Bis(trifluoromethyl)benzylamine
Punto de fusión 54°C to 60°C
Punto de ebullición 81°C to 83°C (13 mmHg)
Temperatura de inflamación 78°C (172°F)
Cantidad 1 g
Número UN UN3259
Beilstein 7209267
Sensibilidad Air Sensitive
Formula Weight (peso de la fórmula) 243.15
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'