Learn More
Alfa Aesar™ 3,4'-Diisopropilbifenilo, 94 %
Catalog Number p-7029878
Click to view available options
Cantidad:
5 g
25 g
100 g
61434-46-6
C18H22
238.37
MFCD00043539
LHNUPUGVRFQTLK-UHFFFAOYSA-N
3,4'-diisopropylbiphenyl, 3,4'-di-iso-propylbiphenyl, 3,4'-diisopropyl-1,1'-biphenyl, unii-gf449ran44, 1,1'-biphenyl,3,4'-bis 1-methylethyl, 1-methylethyl-3-4-methylethyl phenyl benzene, 1-isopropyl-3-4-isopropylphenyl benzene, 3,4-di-iso-propylbiphenyl, 1,1'-biphenyl, 3,4'-diisopropyl, 3,4'-diisopropyl-1,1'-biphenyl #
123379
1-propan-2-il-3-(4-propan-2-ilfenil)benceno
CC(C)C1=CC=C(C=C1)C1=CC=CC(=C1)C(C)C
61434-46-6
C18H22
238.37
MFCD00043539
LHNUPUGVRFQTLK-UHFFFAOYSA-N
3,4'-diisopropylbiphenyl, 3,4'-di-iso-propylbiphenyl, 3,4'-diisopropyl-1,1'-biphenyl, unii-gf449ran44, 1,1'-biphenyl,3,4'-bis 1-methylethyl, 1-methylethyl-3-4-methylethyl phenyl benzene, 1-isopropyl-3-4-isopropylphenyl benzene, 3,4-di-iso-propylbiphenyl, 1,1'-biphenyl, 3,4'-diisopropyl, 3,4'-diisopropyl-1,1'-biphenyl #
123379
1-propan-2-il-3-(4-propan-2-ilfenil)benceno
CC(C)C1=CC=C(C=C1)C1=CC=CC(=C1)C(C)C