Learn More
3,4-Dicloroanisol, 98 %, Thermo Scientific™
Catalog Number p-7023562
Click to view available options
Cantidad:
5 g
25 g
100 g
36404-30-5
C7H6Cl2O
177.024
MFCD00070785
VISJRVXHPNMYRH-UHFFFAOYSA-N
3,4-dichloroanisole, benzene, 1,2-dichloro-4-methoxy, 3,4-dichloromethoxybenzene, 1,2-dichloro-4-methyxybenzene, 3,4-dichloroanizole, pubchem3683, acmc-1ajco, 4,5-dichlorophenyl methyl ether, 3,4-dichlorophenol, methyl ether
37438
1,2-dicloro-4-metoxibenceno
COC1=CC(=C(C=C1)Cl)Cl
36404-30-5
C7H6Cl2O
177.024
MFCD00070785
VISJRVXHPNMYRH-UHFFFAOYSA-N
3,4-dichloroanisole, benzene, 1,2-dichloro-4-methoxy, 3,4-dichloromethoxybenzene, 1,2-dichloro-4-methyxybenzene, 3,4-dichloroanizole, pubchem3683, acmc-1ajco, 4,5-dichlorophenyl methyl ether, 3,4-dichlorophenol, methyl ether
37438
1,2-dicloro-4-metoxibenceno
COC1=CC(=C(C=C1)Cl)Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3,4-Dichloroanisole
Punto de fusión -8°C
Densidad 1.33
Punto de ebullición 200°C to 204°C
Temperatura de inflamación >110°C (230°F)
Índice de refracción 1.557
Cantidad 5 g
Beilstein 775080
Formula Weight (peso de la fórmula) 177.03
Porcentaje de pureza 98%