missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Tribromopiridina, 97 %, Thermo Scientific™

Catalog Number p-7030667
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
2457-48-9
C5H2Br3N
315.79
MFCD01646053
CWYVUHWKGWQPLP-UHFFFAOYSA-N
pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine
541462
3,4,5-tribromopyridine
BrC1=CN=CC(Br)=C1Br
Este artículo no se puede devolver. Vea la política de devoluciones
2457-48-9
C5H2Br3N
315.79
MFCD01646053
CWYVUHWKGWQPLP-UHFFFAOYSA-N
pubchem13500, 3,4,5-tribromo-pyridine, acmc-20a7i7, pyridine,3,4,5-tribromo, 3,4,5-tribromopyridine
541462
3,4,5-tribromopyridine
BrC1=CN=CC(Br)=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3,4,5-Tribromopyridine
Punto de fusión 105°C to 109°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 315.79
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'