Learn More
(3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-hydroxyphenyl]-2-phenyl-1-butene, TRC
Product Code. 30359498
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359498 100 mg 100mg
30355602 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30359498 Supplier Toronto Research Chemicals Supplier No. TRCC365754100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-hydroxyphenyl]-2-phenyl-1-butene
CAS 1020719-27-0
Molecular Formula C24H18D5ClO2
Quantity 100 mg
Synonym (E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-(4-hydroxyphenyl)-2-phenyl-1-butene-d5, 4-[1-[4-(2-Chloroethoxy)phenyl]-2-phenyl-1-butenyl-d5]-phenol
InChI Formula InChI=1S/C24H23ClO2/c1-2-23(18-6-4-3-5-7-18)24(19-8-12-21(26)13-9-19)20-10-14-22(15-11-20)27-17-16-25/h3-15,26H,2,16-17H2,1H3/b24-23-/i1D3,2D2
SMILES [2H]C([2H])([2H])C([2H])([2H])\C(=C(/c1ccc(O)cc1)\c2ccc(OCCCl)cc2)\c3ccccc3
IUPAC Name 4-[(Z)-1-[4-(2-chloroethoxy)phenyl]-3,3,4,4,4-pentadeuterio-2-phenylbut-1-enyl]phenol
Molecular Weight (g/mol) 383.92
Formula Weight 383.17