missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3-(2,3,3-trimetil-3H-indol-5-il)propiónico, 96 %, Thermo Scientific™

Catalog Number p-7031831
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
1072069-91-0
C14H17NO2
231.295
MFCD12922972
QIMIOCZMNPBLPO-UHFFFAOYSA-N
3-2,3,3-trimethyl-3h-indol-5-yl propanoic acid, 3-2,3,3-trimethylindol-5-yl propanoic acid, 3-2,3,3-trimethyl-3h-indol-5-yl propionic acid
45497368
ácido 3-(2,3,3-trimetilindol-5-il)propanoico
CC1=NC2=C(C1(C)C)C=C(C=C2)CCC(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
1072069-91-0
C14H17NO2
231.295
MFCD12922972
QIMIOCZMNPBLPO-UHFFFAOYSA-N
3-2,3,3-trimethyl-3h-indol-5-yl propanoic acid, 3-2,3,3-trimethylindol-5-yl propanoic acid, 3-2,3,3-trimethyl-3h-indol-5-yl propionic acid
45497368
ácido 3-(2,3,3-trimetilindol-5-il)propanoico
CC1=NC2=C(C1(C)C)C=C(C=C2)CCC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(2,3,3-Trimethyl-3H-indol-5-yl)propionic acid
Punto de fusión 145°C to 146°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 231.29
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'