missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)acrylic acid, 98%

Product Code. 11468406
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468406 50 g 50g
11458406 10 g 10g
11478406 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468406 Supplier Thermo Scientific Alfa Aesar Supplier No. A15422.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13026-12-5 | C13H10O2 | 198.22 g/mol

3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(1-Naphthyl)acrylic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13026-12-5
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014317,MFCD00014317
InChI Key WPXMLUUYWNHQOR-CMDGGOBGSA-N
Synonym naphthalene-1-acrylic acid, z-3-1-naphthyl acrylic acid, z-3-naphthalen-1-yl-acrylic acid, z-3-naphthalen-1-yl acrylic acid, 2z-3-naphthalen-1-yl prop-2-enoic acid
PubChem CID 12158295
SMILES OC(=O)\C=C\C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 210°C to 212°C
Quantity 50 g
Beilstein 1868617
Solubility Information Slightly soluble in water.
IUPAC Name (Z)-3-naphthalen-1-ylprop-2-enoic acid
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3-(1-Naphthyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.