missing translation for 'onlineSavingsMsg'
Learn More

Ácido 3-(1-cianoetil)benzoico, 98 %, Thermo Scientific™

Catalog Number p-7058110
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
5537-71-3
C10H9NO2
175.19
MFCD00002520
IRYIYPWRXROPSX-UHFFFAOYNA-N
3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid
98370
CHEBI:85131
ácido 3-(1-cianoetil)benzoico
CC(C#N)C1=CC=CC(=C1)C(O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
5537-71-3
C10H9NO2
175.19
MFCD00002520
IRYIYPWRXROPSX-UHFFFAOYNA-N
3-1-cyanoethyl benzoic acid, m-1-cyanoethyl benzoic acid, benzoic acid, 3-1-cyanoethyl, zlchem 504, acmc-1b01z, 3-1-cyanethyl benzoesaeure, 3-1-cyanoethyl benzoicacid, 3-l-cyanoethyl benzoic acid, 3-1-cyanethyl benzoic acid, meta-1-cyanoethyl benzoic acid
98370
CHEBI:85131
ácido 3-(1-cianoetil)benzoico
CC(C#N)C1=CC=CC(=C1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 3-(1-Cyanoethyl)benzoic acid
Punto de fusión 145°C to 148°C
Cantidad 1 g
Formula Weight (peso de la fórmula) 175.18
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'