missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-(-)-2,4-bis(difenilfosfina)pentano, 99 %, Thermo Scientific Chemicals

Catalog Number p-7050052
Click to view available options
Cantidad:
250 mg
Este artículo no se puede devolver. Vea la política de devoluciones
77876-39-2
C29H30P2
440.51
MFCD00058923
CTYPJIUQROQJBG-ISIPKWIHNA-N
2s,4s---2,4-bis diphenylphosphino pentane, s,s-bdpp, 2s,4s-pentane-2,4-diylbis diphenylphosphine, phosphine, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenyl, s,s-2,4-bis diphenylphosphino pentane, 2s,4s-2,4-bis diphenylphosphino pentane, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenylphosphine, 2s,4s-4-diphenylphosphanylpentan-2-yl-diphenylphosphane, 2s,4s-4-diphenylphosphanyl pentan-2-yl diphenylphosphane
2734567
C[C@@H](C[C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
77876-39-2
C29H30P2
440.51
MFCD00058923
CTYPJIUQROQJBG-ISIPKWIHNA-N
2s,4s---2,4-bis diphenylphosphino pentane, s,s-bdpp, 2s,4s-pentane-2,4-diylbis diphenylphosphine, phosphine, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenyl, s,s-2,4-bis diphenylphosphino pentane, 2s,4s-2,4-bis diphenylphosphino pentane, 1s,3s-1,3-dimethyl-1,3-propanediyl bis diphenylphosphine, 2s,4s-4-diphenylphosphanylpentan-2-yl-diphenylphosphane, 2s,4s-4-diphenylphosphanyl pentan-2-yl diphenylphosphane
2734567
C[C@@H](C[C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (2S,4S)-(-)-2,4-Bis(diphenylphosphino)pentane
Cantidad 250 mg
Nombre IUPAC [(2S,4S)-4-difenilfosfanilpentan-2-il]-difenilfosfano
Formula Weight (peso de la fórmula) 440.5
Porcentaje de pureza 99%
Forma física cristalino/a/s

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'