Learn More
(2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl Ester 2-Methyl Ester, TRC
Product Code. 30396718
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30396718 5 g 5g
1 options
This item is not returnable. View return policy
Product Code. 30396718 Supplier Toronto Research Chemicals Supplier No. TRCH6565605G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-butyl ester 2-methyl ester
CAS 74844-91-0
Molecular Formula C11 H19 N O5
Quantity 5 g
Synonym 1,2-Pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S,4R)-, 1,2-Pyrrolidinedicarboxylic acid, 4-hydroxy-, 1-(1,1-dimethylethyl) 2-methyl ester, (2S-trans)-, (2S,4R)-1-tert-Butoxycarbonyl-2-(methoxycarbonyl)-4-hydroxypyrrolidine, (2S,4R)-1-tert-Butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid methyl ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, (2S,4R)-Methyl 1-(tert-butyloxycarbonyl)-4-hydroxypyrrolidine-2-carboxylate, 1-tert-Butyl 2-methyl (2S,4R)-4-hydroxy-1,2-pyrrolidinedicarboxylate, 1-tert-Butyl 2-methyl 4(R)-hydroxypyrrolidine-1,2(S)-dicarboxylate, Methyl (2S,4R)-1-(tert-butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylate, Methyl (2S,4R)-N-tert-Butoxycarbonyl-4-hydroxy-2-pyrrolidinecarboxylate, Methyl (2S,4R)-N-tert-butoxycarbonyl-4-hydroxy-2-pyrrolidinecarboxylate, Methyl (4R)-N-Boc-4-hydroxy-L-prolinate, Methyl N-tert-butoxycarbonyl-L-hydroxyprolinate, Methyl-(N-Boc-4R-hydroxypyrrolidine)-2-carboxylate, N-(tert-Butoxycarbonyl)-(2S,4R)-4-hydroxyproline methyl ester, N-(tert-Butoxycarbonyl)-(2S,4R)-4-hydroxyproline methyl ester, N-Boc-4-hydroxy-L-proline methyl ester, N-Boc-trans-4-hydroxyproline methyl ester, N-tert-Butoxycarbonyl-L-hydroxyproline methyl ester, tert-Butyl (2S,4R)-2-(methoxycarbonyl)-4-hydroxypyrrolidine-1-carboxylate, tert-Butyl methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
InChI Formula InChI=1S/C11H19NO5/c1-11(2,3)17-10(15)12-6-7(13)5-8(12)9(14)16-4/h7-8,13H,5-6H2,1-4H3/t7-,8+/m1/s1
SMILES COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol) 245.27
Formula Weight 245.1263