missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-2-(Boc-amino)-3-benciloxi-1-butanol, 97 %, Thermo Scientific™

Catalog Number p-7034865
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
133565-43-2
C16H25NO4
295.38
MFCD00235948
RXDBGDZAKNELGW-UHFFFAOYNA-N
tert-butyl 2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate, boc-thr bzl-ol, boc-threoninol bzl, 2r,3r-2-boc-amino-3-benzyloxy-1-butanol, tert-butyl n-2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate, ambotzbal1043, n-boc-l-thr bzl-ol, n-boc-o-benzyl-l-threoninol, n-boc-2r,3r-2-amino-3-benzyloxy-1-butanol
11109205
N-[(2R,3R)-1-hidroxi-3-fenilmetoxibutan-2-il]carbamato de terc-butilo
CC(OCC1=CC=CC=C1)C(CO)NC(=O)OC(C)(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
133565-43-2
C16H25NO4
295.38
MFCD00235948
RXDBGDZAKNELGW-UHFFFAOYNA-N
tert-butyl 2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate, boc-thr bzl-ol, boc-threoninol bzl, 2r,3r-2-boc-amino-3-benzyloxy-1-butanol, tert-butyl n-2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate, ambotzbal1043, n-boc-l-thr bzl-ol, n-boc-o-benzyl-l-threoninol, n-boc-2r,3r-2-amino-3-benzyloxy-1-butanol
11109205
N-[(2R,3R)-1-hidroxi-3-fenilmetoxibutan-2-il]carbamato de terc-butilo
CC(OCC1=CC=CC=C1)C(CO)NC(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material (2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol
Cantidad 250 mg
Formula Weight (peso de la fórmula) 295.38
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'