Learn More
(2E)-3-[[(4-Methylphenyl)sulfonyl]oxy]-2-butenoic Acid Methyl Ester, TRC
Product Code. 30469560
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30469560 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30469560 Supplier Toronto Research Chemicals Supplier No. TRCM3274501G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2E)-3-(((4-Methylphenyl)sulfonyl)oxy)-2-butenoic Acid Methyl Ester
CAS 1029612-16-5
Molecular Formula C12H14O5S
Quantity 1 g
InChI Formula InChI=1S/C12H14O5S/c1-9-4-6-11(7-5-9)18(14,15)17-10(2)8-12(13)16-3/h4-8H,1-3H3/b10-8+
SMILES CC1=CC=C(C=C1)S(=O)(=O)O/C(=C/C(=O)OC)/C
Molecular Weight (g/mol) 270.3