missing translation for 'onlineSavingsMsg'
Learn More

2A Peptide Antibody (3H4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-59627AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214292

  • 561.54€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

2A Peptide Monoclonal specifically detects 2A Peptide in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

2A Peptide
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
Mouse
0.1 mL
This 2A peptide antibody is specific for T2A and P2A tagged proteins. Specificity to F2A and E2A proteins has not been tested.
Store at 4°C in the dark.
Western Blot, Immunocytochemistry, Immunofluorescence
3H4
Western Blot, Immunocytochemistry/Immunofluorescence
2A cleavage peptide, 2A peptide, 2A peptide tag, 2A sequence, Anti- MDV 2A-peptide, anti-2A peptide, anti-2A sequence, anti-2A-like peptide, antibody for 2A-containing recombinant proteins, anti-P2A tagged proteins, anti-P2A peptide, anti-T2A tagged proteins, anti-T2A peptide, anti-Thosea asigna virus 2A, FMDV 2A-peptide, peptide 2A sequences, self-cleaving′2A peptide2a pept, Thosea asigna virus 2A
KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG)
Primary
Non-species specific
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales