Learn More
Code Corporation™ 2700 Inductive Charging Station Base Mount
Product Code. 17900002 Shop All Brady Products
Change view
Click to view available options
Connection Type:
RS-232, USB
Unit Size:
Each
Product Code. Connection Type Unit Size
17900002 RS-232, USB Each
1 options
This item is not returnable. View return policy
Product Code. 17900002 Supplier Code Corporation™ Supplier No. 176690
  • ONLINE EXCLUSIVE PRICE
    57.95€
    54.70€ / paquete de 1
    Ahorro 3.25€
    5% Descuento
Fecha estimada de envoi: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Keep your CR2700 Inductive Charging Station steady and right where you need it with this sturdy base mount.
  • Keep your CR2700 Inductive Charging Station steady and right where you need it with this sturdy base mount.
  • For use with CR2700 Inductive Charging Station.
Specifications
Description Code Reader™ 2700 Inductive Charging Station Base Mount
Height (Metric) 21.08 mm
Height (English) 0.83 in.
Width (Metric) 93.98 mm
Width (English) 3.7 in.
Depth (Metric) 121.92 mm
Depth (English) 4.8 in.
For Use With (Application) Asset Tracking, General Identification Healthcare, Laboratory, Manufacturing, Pharmaceutical
Quantity 1 Ea.
Type Charging Station Base Mount
Connection Type RS-232, USB
Data Memory None
Display Type None Included
Power Supply USB or RS-232
Resolution No Display Included
Alternate Part Number CRA-MB6-C286