Learn More
(+)-20-Hydroxyecdysone, TRC
Product Code. 30012417
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012417 10 g 10g
30006676 1 g 1g
30004085 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012417 Supplier Toronto Research Chemicals Supplier No. TRCH91875010G
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5289-74-7
Molecular Formula C27 H44 O7
Molecular Weight (g/mol) 480.63
Synonym Cholest-7-en-6-one, 2,3,14,20,22,25-hexahydroxy-, (2β,3β,5β,22R)-, 5β-Cholest-7-en-6-one, 2β,3β,14,20,22,25-hexahydroxy-, (22R)- (8CI), 5β-Cholest-7-en-6-one, 2β,3β,14,20,22β,25-hexahydroxy- (7CI), (+)-20-Hydroxyecdysone, (+)-Ecdysterone, (2β,3β,5β,22R)-2,3,14,20,22,25-Hexahydroxycholest-7-en-6-one, 20-Hydroxy-α-ecdysone, 20-Hydroxyecdysone, 20R-Hydroxyecdysone, 2β,3β,14α,20β,22α,25-Hexahydroxycholest-7-en-6-one, Commisterone, Crustecdyson, Crustecdysone, Ecdysone, Ecdysten, Ecdysteron, Ecdysterone, Ekdisten, Isoinokosterone, Polypodin A, Polypodin C, Polypodine A, Polypodine C, THE 7, Viticosterone, β-Ecdysone, β-Ecdysterone, β-Ecydisone
IUPAC Name (2S,3R,5R,9R,10R,13R,14S,17S)-2,3,14-trihydroxy-10,13-dimethyl-17-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-2,3,4,5,9,11,12,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-6-one
SMILES CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 10 g
InChI Formula InChI=1S/C27H44O7/c1-23(2,32)9-8-22(31)26(5,33)21-7-11-27(34)16-12-18(28)17-13-19(29)20(30)14-24(17,3)15(16)6-10-25(21,27)4/h12,15,17,19-22,29-34H,6-11,13-14H2,1-5H3/t15-,17-,19+,20-,21-,22+,24+,25+,26+,27+/m0/s1
Formula Weight 480.3087
Chemical Name or Material beta-Ecdysone