missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluorometil)fenotiazina, 98 %

Catalog Number p-7025447
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
92-30-8
C13H8F3NS
267.269
MFCD00005018
RKGYJVASTMCSHZ-UHFFFAOYSA-N
2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference
7082
2-(trifluorometil)-10H-fenotiazina
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
92-30-8
C13H8F3NS
267.269
MFCD00005018
RKGYJVASTMCSHZ-UHFFFAOYSA-N
2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference
7082
2-(trifluorometil)-10H-fenotiazina
C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-(Trifluoromethyl)phenothiazine
Punto de fusión 188°C to 190°C
Cantidad 5 g
Beilstein 226580
Formula Weight (peso de la fórmula) 267.27
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'