Learn More
2-(Trifluoromethyl)benzyl chloride, 98+%, Thermo Scientific™
Product Code. 11489087
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489087 25 g 25g
11479087 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489087 Supplier Thermo Scientific Alfa Aesar Supplier No. B20648.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21742-00-7
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
MDL Number MFCD00009920
InChI Key BBXDMCQDLOCXRA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzyl chloride, 1-chloromethyl-2-trifluoromethyl benzene, 2-trifluoromethyl benzylchloride, 2-trifluoromethylbenzyl chloride, o-trifluoromethylbenzyl chloride, benzene, 1-chloromethyl-2-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-o-xylene, g1r bxfff, pubchem4692, acmc-209fnp
PubChem CID 89028
IUPAC Name 1-(chloromethyl)-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)C(F)(F)F
Specifications
Density 1.337
Boiling Point 67°C to 68°C (12 mmHg)
Flash Point 70°C (158°F)
Odor Pungent
Refractive Index 1.47
Quantity 25 g
UN Number UN3265
Beilstein 389380
Formula Weight 194.59
Percent Purity ≥98%
Chemical Name or Material 2-(Trifluoromethyl)benzyl chloride