missing translation for 'onlineSavingsMsg'
Learn More

Ácido 2-(trifluorometoxi)benzoico, 98 %, Thermo Scientific™

Catalog Number p-7081458
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
1979-29-9
C8H4F3O3
205.11
MFCD00052325
JMYSPFGUBNENSE-UHFFFAOYSA-M
2-trifluoromethoxy benzoic acid, o-trifluoromethoxybenzoic acid, alpha,alpha,alpha-trifluoro-o-anisic acid, 2-trifluoromethoxy-benzoic acid, rarechem al bo 0449, pubchem4587, acmc-209f2a, ksc494o7t
2777223
ácido 2-(trifluorometoxi)benzoico
[O-]C(=O)C1=CC=CC=C1OC(F)(F)F
Este artículo no se puede devolver. Vea la política de devoluciones
1979-29-9
C8H4F3O3
205.11
MFCD00052325
JMYSPFGUBNENSE-UHFFFAOYSA-M
2-trifluoromethoxy benzoic acid, o-trifluoromethoxybenzoic acid, alpha,alpha,alpha-trifluoro-o-anisic acid, 2-trifluoromethoxy-benzoic acid, rarechem al bo 0449, pubchem4587, acmc-209f2a, ksc494o7t
2777223
ácido 2-(trifluorometoxi)benzoico
[O-]C(=O)C1=CC=CC=C1OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2- (Trifluoromethoxy)benzoic acid
Punto de fusión 78°C to 80°C
Cantidad 1 g
Beilstein 2645481
Formula Weight (peso de la fórmula) 206.12
Porcentaje de pureza 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'