Learn More
2-tert-Butyl-4-Hydroxyanisole, TRC
Product Code. 30138054
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138054 500 mg 500mg
30134431 5 g 5g
17991855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138054 Supplier Toronto Research Chemicals Supplier No. TRCB699300500MG
 Restricted Item
Fecha estimada de envoi: 20-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88-32-4
Molecular Formula C11 H16 O2
Molecular Weight (g/mol) 180.24
Synonym 3-(1,1-Dimethylethyl)-4-methoxyphenol, 3-tert-Butyl-4-methoxyphenol, 2-(1,1-Dimethylethyl)-4-hydroxyanisole, 2-tert-Butyl-4-hydroxyanisole, 2-tert-Butyl-p-hydroxyanisole, 4-Methoxy-3-(1,1-dimethylethyl)phenol, 4-Methoxy-3-tert-butylphenol, tert-Butyl-3-methoxy-4-phenol
IUPAC Name 3-tert-butyl-4-methoxyphenol
SMILES COc1ccc(O)cc1C(C)(C)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C11H16O2/c1-11(2,3)9-7-8(12)5-6-10(9)13-4/h5-7,12H,1-4H3
Formula Weight 180.115
Chemical Name or Material 2-tert-Butyl-4-hydroxyanisole