Learn More
2-sec-Butyl-d5-4-{4-[4-(4-hydroxy-phenyl)-piperazin-1-yl]-phenyl}-2,4-dihydro-[1,2,4]-triazol-3-one, TRC
Product Code. 30332242
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30332242 5 mg 5mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30332242 Supplier Toronto Research Chemicals Supplier No. TRCB6920275MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-sec-Butyl-d5-4-{4-[4-(4-hydroxy-phenyl)-piperazin-1-yl]-ph
CAS 1020719-21-4
Molecular Formula C22 2H5 H22 N5 O2
Quantity 5 mg
InChI Formula InChI=1S/C22H27N5O2/c1-3-17(2)27-22(29)26(16-23-27)20-6-4-18(5-7-20)24-12-14-25(15-13-24)19-8-10-21(28)11-9-19/h4-11,16-17,28H,3,12-15H2,1-2H3/i1D3,3D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C(C)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(O)cc4
IUPAC Name 4-[4-[4-(4-hydroxyphenyl)piperazin-1-yl]phenyl]-2-(3,3,4,4,4-pentadeuteriobutan-2-yl)-1,2,4-triazol-3-one
Molecular Weight (g/mol) 398.51
Formula Weight 398.2479