missing translation for 'onlineSavingsMsg'
Learn More

2-Fenil-2-pentanol, 98 %, Thermo Scientific Chemicals

Catalog Number p-7025403
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
4383-18-0
C11H16O
164.248
MFCD00046592
YBQOTTAEVBHNEJ-UHFFFAOYSA-N
2-phenyl-2-pentanol, 2-phenyl-pentan-2-ol, benzenemethanol, .alpha.-methyl-.alpha.-propyl, methyl phenylbutanol, acmc-1aei8, methyl phenyl n-propyl carbinol
138214
2-Fenilpentan-2-ol
CCCC(C)(C1=CC=CC=C1)O
Este artículo no se puede devolver. Vea la política de devoluciones
4383-18-0
C11H16O
164.248
MFCD00046592
YBQOTTAEVBHNEJ-UHFFFAOYSA-N
2-phenyl-2-pentanol, 2-phenyl-pentan-2-ol, benzenemethanol, .alpha.-methyl-.alpha.-propyl, methyl phenylbutanol, acmc-1aei8, methyl phenyl n-propyl carbinol
138214
2-Fenilpentan-2-ol
CCCC(C)(C1=CC=CC=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Phenyl-2-pentanol
Cantidad 1 g
Formula Weight (peso de la fórmula) 164.25
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'