missing translation for 'onlineSavingsMsg'
Learn More

Alcohol 2-fenoxibencílico, 97 %, Thermo Scientific Chemicals

Catalog Number p-7032004
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
13807-84-6
C13H12O2
200.237
MFCD00017297
VMZBMTWFHYYOIN-UHFFFAOYSA-N
2-phenoxyphenyl methanol, 2-phenoxybenzyl alcohol, o-phenoxybenzyl alcohol, phenoxyphenylmethanol, 2-phenoxybenzylalcohol, 2-phenoxybenzenemethanol, o-phenoxy benzyl alcohol, 2-phenoxy-benzenemethanol, benzenemethanol, phenoxy, 2-phenoxy-phenyl-methanol
3660111
(2-fenoxifenil)metanol
C1=CC=C(C=C1)OC2=CC=CC=C2CO
Este artículo no se puede devolver. Vea la política de devoluciones
13807-84-6
C13H12O2
200.237
MFCD00017297
VMZBMTWFHYYOIN-UHFFFAOYSA-N
2-phenoxyphenyl methanol, 2-phenoxybenzyl alcohol, o-phenoxybenzyl alcohol, phenoxyphenylmethanol, 2-phenoxybenzylalcohol, 2-phenoxybenzenemethanol, o-phenoxy benzyl alcohol, 2-phenoxy-benzenemethanol, benzenemethanol, phenoxy, 2-phenoxy-phenyl-methanol
3660111
(2-fenoxifenil)metanol
C1=CC=C(C=C1)OC2=CC=CC=C2CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Phenoxybenzyl alcohol
Cantidad 250 mg
Formula Weight (peso de la fórmula) 200.24
Porcentaje de pureza 97%

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'