Learn More
2-Nitrobenzhidrazida, 98 %, Thermo Scientific Chemicals
Catalog Number p-7025695
Click to view available options
Cantidad:
5 g
25 g
100 g
606-26-8
C7H7N3O3
181.151
MFCD00007598
LYGGDXLOJMNFBV-UHFFFAOYSA-N
2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
3781218
2-nitrobenzohidrazida
C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]
606-26-8
C7H7N3O3
181.151
MFCD00007598
LYGGDXLOJMNFBV-UHFFFAOYSA-N
2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
3781218
2-nitrobenzohidrazida
C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
La reacción de 2,3-dicloro-1,4-naftoquinona con p-nitrobenzhidrazida genera bis-(p-nitrobenzhidrazino)-1,4-naptquinona de producto disustituido.
Solubilidad
Soluble en metanol (casi transparencia).
Notas
Almacenar en un lugar fresco. Mantenga el recipiente bien cerrado en un lugar seco y bien ventilado. Almacenar lejos de los agentes oxidantes fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Nitrobenzhydrazide
Punto de fusión 119°C to 123°C
Cantidad 5 g
Beilstein 394754
Información de solubilidad Soluble in methanol(almost transparency).
Formula Weight (peso de la fórmula) 181.15
Porcentaje de pureza 98%