Learn More
2-Nitrobenzaldehyde-d4, TRC
Product Code. 30469582
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469582 5 mg 5mg
30468768 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30469582 Supplier Toronto Research Chemicals Supplier No. TRCN4921275MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Nitrobenzaldehyde-d4
CAS 1020718-69-7
Molecular Formula C7 2H4 H N O3
Quantity 5 mg
Synonym 2-Nitrobenzaldehyde D4
InChI Formula InChI=1S/C7H5NO3/c9-5-6-3-1-2-4-7(6)8(10)11/h1-5H/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c(c(C=O)c1[2H])[N+](=O)[O-]
IUPAC Name 2,3,4,5-tetradeuterio-6-nitrobenzaldehyde
Molecular Weight (g/mol) 155.14
Formula Weight 155.0521