missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-N-(2-nitrofenil)bencenosulfonamida, 97 %, Thermo Scientific™

Catalog Number p-7040423
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
331256-09-8
C12H9N3O6S
323.279
MFCD00406329
NGHQCODQNBAKLD-UHFFFAOYSA-N
2-nitro-n-2-nitrophenyl benzenesulfonamide, n-2-nitrophenylsulfonyl-2-nitroaniline, 2-nitrophenyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-2-nitro-phenyl-benzenesulfonamide
2827773
2-nitro-N-(2-nitrofenil)bencenosulfonamida
C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
331256-09-8
C12H9N3O6S
323.279
MFCD00406329
NGHQCODQNBAKLD-UHFFFAOYSA-N
2-nitro-n-2-nitrophenyl benzenesulfonamide, n-2-nitrophenylsulfonyl-2-nitroaniline, 2-nitrophenyl 2-nitrophenyl sulfonyl amine, 2-nitro-n-2-nitro-phenyl-benzenesulfonamide
2827773
2-nitro-N-(2-nitrofenil)bencenosulfonamida
C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-])[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Nitro-N-(2-nitrophenyl)benzenesulfonamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 323.28
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'