Learn More
2-(n-Propylthio)ethanol, 98%, Thermo Scientific™
Product Code. 11343216
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343216 5 g 5g
11353216 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343216 Supplier Thermo Scientific Alfa Aesar Supplier No. L12528.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22812-90-4
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00014041
InChI Key KCWWXXYQPUDKBX-UHFFFAOYSA-N
Synonym 2-propylthio ethanol, 2-hydroxyethyl propyl sulfide, 2-n-propylthio ethanol, 2-hydroxyethyl n-propyl sulfide, 2-propylsulfanyl ethanol, 2-propylthioethan-1-ol, acmc-209fyz, ethanol, 2-propylthio, 2-propylsulfanyl ethanol #, 2-hydroxyethyln-propylsulfide
PubChem CID 89850
IUPAC Name 2-propylsulfanylethanol
SMILES CCCSCCO
Specifications
Density 0.99
Boiling Point 199°C to 200°C
Flash Point 98°C (208°F)
Odor Unpleasant
Linear Formula CH3(CH2)2S(CH2)2OH
Refractive Index 1.4815
Quantity 5 g
UN Number UN1760
Beilstein 1731984
Formula Weight 120.21
Percent Purity 98%
Chemical Name or Material 2-(n-Propylthio)ethanol