Learn More
2-Methylthiophenothiazine, Mikromol™
Product Code. 30464061
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464061 4 x 25 mg 100mg
30051908 25 mg 25mg
30051315 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464061 Supplier Mikromol Supplier No. MM0116.0500254AMP
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methylthiophenothiazine, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methylthiophenothiazine
CAS 8-5-7643
Analyte or Component Names 2-Methylthiophenothiazine
Shipping Condition Room Temperature
Molecular Formula C13 H11 N S2
Certifications/Compliance ISO 17025
Synonym 2-Methylthiophenothiazine
InChI Formula InChI=1S/C13H11NS2/c1-15-9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)16-13/h2-8,14H,1H3
Solution Type Neat
SMILES CSc1ccc2Sc3ccccc3Nc2c1
IUPAC Name 2-methylsulfanyl-10H-phenothiazine
Molecular Weight (g/mol) 245.36
Quantity 4 x 25 mg
Formula Weight 245.0333 g/mol