Learn More
2-Methyl-7-methylamino-d3-8-nitro-quinoxaline, TRC
Product Code. 30465020
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30465020 100 mg 100mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30465020 Supplier Toronto Research Chemicals Supplier No. TRCM322407100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-7-methylamino-d3-8-nitro-quinoxaline
CAS 1020718-63-1
Molecular Formula C10 D3 H7 N4 O2
Quantity 100 mg
InChI Formula InChI=1S/C10H10N4O2/c1-6-5-12-7-3-4-8(11-2)10(14(15)16)9(7)13-6/h3-5,11H,1-2H3/i2D3
SMILES [2H]C([2H])([2H])Nc1ccc2ncc(C)nc2c1[N+](=O)[O-]
IUPAC Name 3-methyl-5-nitro-N-(trideuteriomethyl)quinoxalin-6-amine
Molecular Weight (g/mol) 221.23
Formula Weight 221.0992