missing translation for 'onlineSavingsMsg'
Learn More

2-Metil-5-(trifluorometoxi)benzaldehído, 96 %, Thermo Scientific™

Catalog Number p-7032358
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
195622-40-3
C9H7F3O2
204.148
MFCD18399706
JTYVBDVMQINBPT-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzaldehyde
59916918
2-metil-5-(trifluorometoxi)benzaldehído
CC1=C(C=C(C=C1)OC(F)(F)F)C=O
Este artículo no se puede devolver. Vea la política de devoluciones
195622-40-3
C9H7F3O2
204.148
MFCD18399706
JTYVBDVMQINBPT-UHFFFAOYSA-N
2-methyl-5-trifluoromethoxy benzaldehyde
59916918
2-metil-5-(trifluorometoxi)benzaldehído
CC1=C(C=C(C=C1)OC(F)(F)F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Methyl-5-(trifluoromethoxy)benzaldehyde
Cantidad 1 g
Sensibilidad Air sensitive
Formula Weight (peso de la fórmula) 204.15
Porcentaje de pureza 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'