Learn More
2-Methyl-5-nitroimidazole, TRC
Product Code. 17993515
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993515 100 g 100g
17921405 10 g 10g
17962495 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993515 Supplier Toronto Research Chemicals Supplier No. TRCM323765100G
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 696-23-1
Molecular Formula C4 H5 N3 O2
IUPAC Name 2-methyl-4-nitro-1H-imidazole
SMILES Cc1nc(c[nH]1)[N+](=O)[O-]
Specifications
Quantity 100 g
Synonym 2-Methyl-4-nitroimidazole, 2-Methyl-5-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole, Metronidazole Benzoate Imp. B (EP), Tinidazole Imp. A (EP), Metronidazole Imp. A (EP), 1H-Imidazole, 2-methyl-5-nitro-, 1H-Imidazole, 2-methyl-4-nitro- (9CI), Imidazole, 2-methyl-4(or 5)-nitro- (7CI), Imidazole, 2-methyl-4-nitro- (8CI), 2-Methyl-4-nitro-1H-imidazole, 4-Nitro-2-methylimidazole, L 581490, Menidazole, RP 8532
InChI Formula InChI=1S/C4H5N3O2/c1-3-5-2-4(6-3)7(8)9/h2H,1H3,(H,5,6)
Molecular Weight (g/mol) 127.1
Formula Weight 127.0382
Percent Purity >95%
Chemical Name or Material 2-Methyl-4-nitroimidazole
Grade HPLC