Learn More
2-Methyl-3-trifluoromethylaniline, TRC
Product Code. 30132793
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132793 5 g 5g
30012544 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132793 Supplier Toronto Research Chemicals Supplier No. TRCM3313455G
 Restricted Item
Fecha estimada de envoi: 19-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54396-44-0
Molecular Formula C8 H8 F3 N
Molecular Weight (g/mol) 175.15
Synonym 2-Amino-6-(trifluoromethyl)toluene, 2-Methyl-3-(trifluoromethyl)aniline, 2-Methyl-3-(trifluoromethyl)benzenamine, 2-Methyl-3-(trifluoromethyl)phenylamine, 2-Methyl-3-aminobenzotrifluoride, 3-(Trifluoromethyl)-2-methylaniline, 3-(Trifluoromethyl)-2-methylbenzenamine
IUPAC Name 2-methyl-3-(trifluoromethyl)aniline
SMILES Cc1c(N)cccc1C(F)(F)F
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H8F3N/c1-5-6(8(9,10)11)3-2-4-7(5)12/h2-4H,12H2,1H3
Formula Weight 175.0609
Chemical Name or Material 2-methyl-3-(trifluoromethyl)aniline