missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-Propanol, ≥99.0%, ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Cantidad:
500 mL
1 L
2.5 L
Envase:
Glass Bottle
Descripción
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Especificaciones
Nombre del producto químico o material | 2-Methyl-1-Propanol |
Punto de fusión | -108°C |
Densidad | 0.803g/mL at 25°C |
Punto de ebullición | 108°C |
Temperatura de inflamación | 28°C (82.4°F) |
Punto de ignición | 801°F |
Envase | Glass Bottle |
Fórmula lineal | (CH3)2CHCH2OH |
Cantidad | 2.5 L |
Número UN | UN1212 |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'