missing translation for 'onlineSavingsMsg'
Learn More

Éster de pinacol de ácido 2-metoxipiridina-3-borónico, 97 %, Thermo Scientific Chemicals

Catalog Number p-7033051
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
532391-31-4
C12H18BNO3
235.09
MFCD05663507
JEJYNUCGRAHMDO-UHFFFAOYSA-N
2-methoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-methoxypyridine-3-boronic acid pinacol ester, 2-methoxypyridin-3-ylboronic acid pinacol ester, 2-methoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pubchem16545, ksc498e8t, 2-methoxypyridine-3-boronic acid, pinacol ester, 2-methoxypyridin-3-yl boronic acid pinacol ester
17750166
2-metoxi-3-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina
COC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1
Este artículo no se puede devolver. Vea la política de devoluciones
532391-31-4
C12H18BNO3
235.09
MFCD05663507
JEJYNUCGRAHMDO-UHFFFAOYSA-N
2-methoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-methoxypyridine-3-boronic acid pinacol ester, 2-methoxypyridin-3-ylboronic acid pinacol ester, 2-methoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pubchem16545, ksc498e8t, 2-methoxypyridine-3-boronic acid, pinacol ester, 2-methoxypyridin-3-yl boronic acid pinacol ester
17750166
2-metoxi-3-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)piridina
COC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2-Methoxypyridine-3-boronic acid pinacol ester
Cantidad 250 mg
Formula Weight (peso de la fórmula) 235.09
Porcentaje de pureza 97%

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'